Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUMO2/SUMO3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA520959

Catalog No. PIPA520959


Add to cart

Description

Description

A suggested positive control is rat liver tissue lysate. PA5-20959 can be used with blocking peptide PEP-1073.

Small ubiquitin-like modifiers (SUMOs) are a family of small, related proteins (Sumo-1/2/3/4) that can be enzymatically attached to a target protein by a post-translational modification process termed sumoylation, which is a major regulator of protein function in cellular processes such as nuclear transport, transcriptional regulation, apoptosis and protein stability. All SUMO proteins localize to the nucleus and are covalently conjugated, affecting protein structure, function and interactions. SUMO2 and 3 are 96% identical and are more mobile within nucleus relative to SUMO1. Specific functional differences between SUMO1 and SUMO2 and 3 remain to be identified.
Specifications

Specifications

SUMO2/SUMO3
Polyclonal
Unconjugated
SUMO2
Liquid
SUMO2, SUMO3
A 15 amino acid peptide near the carboxy terminus of human SUMO2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P55854, P61956, P61957, Q9Z172, P61959, Q5XIF4
Sumo3, SUMO2, SMT3A, SMT3H1, SUMO-3, Smt3B, SMT3 homolog 1, SMT3 suppressor of mif two 3 homolog 1, SUMO-2, small ubiquitin-related modifier 3, ubiquitin-like protein SMT3A, ubiquitin-like protein SMT3B
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
170930, 20610, 499417, 6612, 6613, 690244
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit