Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUMO2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527636

Catalog No. PIPA527636


Add to cart

Description

Description

PA5-27636 targets SUMO2 in WB applications and shows reactivity with Human and mouse samples. The PA5-27636 immunogen is recombit protein fragment corresponding to a region within amino acids 1 and 84 of Human SUMO2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a protein that is a member of the SUMO (small ubiquitin-like modifier) protein family. It functions in a manner similar to ubiquitin in that it is bound to target proteins as part of a post-translational modification system. However, unlike ubiquitin which targets proteins for degradation, this protein is involved in a variety of cellular processes, such as nuclear transport, transcriptional regulation, apoptosis, and protein stability. It is not active until the last two amino acids of the carboxy-terminus have been cleaved off. Numerous pseudogenes have been reported for this gene. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications

Specifications

SUMO2
Polyclonal
Unconjugated
SUMO2
Liquid
SUMO2
Recombit protein fragment corresponding to a region within amino acids 1 and 84 of Human SUMO2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.56 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P61956, P61957
HSMT3, SMT3B, SMT3H2, SUMO3, Smt3A, SMT3 homolog 2, sentrin 2, small ubiquitin-like modifier 2, small ubiquitin-related modifier 2, ubiquitin-like protein SMT3A, ubiquitin-like protein SMT3B
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
170930, 6613
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit