Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sulfite oxidase, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human SUOX protein.

Manufacturer:  Abnova Corporation H00006821B01

Catalog No. 89-006-309


Add to cart

Description

Description

Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins. [provided by RefSeq

Sequence: MGTLLGLGAVLAYQDHRCRAAQESTHIYTKEEVSSHTSPETGIWVTLGSEVFDVTEFVDLHPGGPSKLMLAAGGPLEPFWALYAVHNQSHVRELLAQYKIGELNPEDKVAPTVETSDPYADDPVRHPALKVNSQRPFNAEPPPELLTENYITPNPIFFTRNHLPVPNLDPDTYRLHVVGAPGGQSLSLSLDDLHNFPRYEITVTLQCAGNRRSEMTQVKEVKGLEWRTGAISTARWAGARLCDVLAQAGHQLCETEAHVCFEGLDSDPTGTAYGASIPLARAMDPEAEVLLAYEMNGQPLPRDHGFPVRVVVPGVVGARHVKWLGRVSVQPEESYSHWQRRDYKGFSPSVDWETVDFDSAPSIQELPVQSAITEPRDGETVESGEVTIKGYAWSGGGRAVIRVDVSLDGGLTWQVAKLDGEEQRPRKAWAWRLWQLKAPVPAGQKELNIVCKAVDDGYNVQPDTVAPIWNLRGVLSNAWHRVHVYVSP
Specifications

Specifications

sulfite oxidase
Unconjugated
No additive
ENST00000356124
SUOX
SUOX (ENSP00000348440, 1 a.a. ~ 488 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
6821
Immunohistochemistry (PFA fixed), Western Blot
Mouse polyclonal antibody raised against a full-length human SUOX protein.
SUOX
Serum
Mouse
50 uL
Endocrine/Metabolism
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit