Learn More
anti-SUR-8, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP148382
Catalog No. NBP148382
Add to cart
Description
Description
SUR-8 Polyclonal antibody specifically detects SUR-8 in Human, Rat samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
SUR-8
LYOPH
Western Blot, ELISA
Antisera
SHOC2
Synthetic peptide derived from N term parts of human protein.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Antibody reacts specifically with 65 kDa human protein.
Human, Rat
ELISA, Western Blot
Unconjugated
Antiserum with No Preservative
KIAA0862FLJ60412, Protein soc-2 homolog, Protein sur-8 homolog, SIAA0862, SOC-2, soc-2 (suppressor of clear, C.elegans) homolog, soc-2 suppressor of clear homolog (C. elegans), SOC2SUR8leucine-rich repeat protein SHOC-2, SUR-8
Rabbit
IgG
0.2 ml
Signal Transduction
Primary
8036
Reconstitute with 0.2 ml distilled water.