Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Survivin, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$155.80 - $240.50

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This Survivin Antibody was developed against full-length recombinant human survivin protein.
Antigen Survivin
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056168T
View Documents
Novus Biologicals
NB10056168SS
0.025 ml
Each for $155.80
Add to cart
 
NB10056168
View Documents
Novus Biologicals
NB10056168
0.05 ml
Each for $240.50
Add to cart
 
Description

Description

Survivin Polyclonal antibody specifically detects Survivin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

Rabbit
This Survivin Antibody was developed against full-length recombinant human survivin protein.
Polyclonal
Primary
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
332
IgG
Survivin
Antisera
RUO
Survivin
Whole antisera with 0.05% Sodium Azide
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit