Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUV420h1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | SUV420h1 |
---|---|
Concentration | 1.16 mg/mL |
Dilution | Flow Cytometry 2-5 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200, Western Blot (Negative) |
Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP197313SS
|
Novus Biologicals
NBP197313SS |
0.025 mL |
Each for $204.00
|
|
|||||
NBP197313
|
Novus Biologicals
NBP197313 |
0.1 mL |
Each for $482.50
|
|
|||||
Description
SUV420h1 Polyclonal specifically detects SUV420h1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative).Specifications
SUV420h1 | |
Flow Cytometry 2-5 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200, Western Blot (Negative) | |
Polyclonal | |
Rabbit | |
Cancer, Epigenetics, Transcription Factors and Regulators | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
C630029K18Rik, CGI85, CGI-85, EC 2.1.1.43, histone-lysine N-methyltransferase SUV420H1, KMT5B, KMT5B Lysine N-methyltransferase 5B, MGC118906, MGC118909, MGC21161, MGC703, Su(var)4-20 homolog 1, Suppressor of variegation 4-20 homolog 1, suppressor of variegation 4-20 homolog 1 (Drosophila), suv4-20h1 | |
KMT5B | |
IgG | |
Affinity Purified |
1.16 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse | |
Q4FZB7, Q3U8K7 | |
51111 | |
A synthetic peptide made to an internal portion of the human SUV420h1 protein (between residues 450-500) [UniProt Q4FZB7] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title