Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUZ12 Mouse anti-Human, Rabbit, Clone: 2AO9, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051317
Description
Use Anti-SUZ12 Antibody, clone 2AO9 (Mouse Monoclonal Antibody) validated in immunoprecipitation, immunocytochemistry to detect SUZ12 also known as ChET 9 protein, Chromatin precipitated E2F target 9 protein, Joined to JAZF1 protein.
Specifications
SUZ12 | |
Monoclonal | |
Unconjugated | |
NP_056170.2 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG |
Immunocytochemistry, Immunoprecipitation | |
2AO9 | |
Purified mouse monoclonal IgG in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
SUZ12, CHET9, JJAZ1 | |
Recombinant protein corresponding to human SUZ12. | |
100 μg | |
Human, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction