Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUZ12 Mouse anti-Human, Clone: 3C1.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 04046
Description
Anti-SUZ12 Antibody, clone 3C1.2 is a high quality Mouse Monoclonal Antibody for the detection of SUZ12 and has been validated in western blotting.Specifications
SUZ12 | |
Monoclonal | |
Unconjugated | |
NM_015355.1 | |
Mouse | |
Protein A purfied | |
Primary | |
−20°C for 24 months. | |
IgG1 κ |
Western Blot | |
3C1.2 | |
100μg of Protein A purified mouse monoclonal IgG1κ in 100μL of 1X PBS, pH 7, 0.1% Azide. | |
SUZ12, KIAA0160, CHET9, JJAZ1 | |
Full length, recombinant human SUZ12. | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only