Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SWAP70, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB10056430SS

 View more versions of this product

Catalog No. NB10056430T


Add to cart

Description

Description

SWAP70 Polyclonal specifically detects SWAP70 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SWAP70
0.5 mg/mL
Western Blot 1-3 ug/ml
Q9UH65
KIAA0640SWAP-70FLJ39540, SWAP switching B-cell complex 70kDa subunit, switch-associated protein 70
Rabbit
IgG
0.025 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Reacts with Human. Predicted to react with Bovine, Chimpanzee, Dog and Mouse.
Western Blot
Unconjugated
PBS containing 0.2% gelatin with 0.05% Sodium Azide
Purified
SWAP70
A portion of amino acids 300-350 of human SWAP-70 was used as the immunogen.
Protein G purified
RUO
Primary
23075
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only