Learn More
SWI6 Rabbit anti-Yeast, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA14977
Catalog No. PIPA14977
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-4977 detects Swi6 from S. pombe samples. PA1-4977 has been successfully used in Western blot and immunoprecimmunoprecipitationitation applications. Western blot analysis of crude extract of S. pombe detects a protein with an apparent molecular size of 53 kD.
Swi6 protein of fission yeast is a functional and structural homolog of HP1 (Heterochromatin Protein 1) of animals and is involved in the formation of heterochromatin structure by binding to centromere, telomere and silent mating-type locus. It is also involved in silencing the genes and sister chromatid cohesion by binding to histone H3 methylated at Lys9 and the cohesin subunit Psc3.
Specifications
Specifications
Swi6
Polyclonal
whole serum with 0.09% sodium azide
P09959
SWI6
Recombit Swi6 from E. coli.
50 μL
-20°C
Primary
850879
ChIP assay, Immunoprecipitation, Western Blot
Unconjugated
SWI6
Liquid
Rabbit
IgG
RUO
Antibody
Polyclonal
Yeast