Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-SWINE IGG(GAMMA) AB FITC  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52300295
Affinity purified polyclonal antibody to Swine IgG heavy chains (gamma) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
Catalog No. 50-668-81