Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-SWINE IGG (H+L) AB HRP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200363
Affinity purified polyclonal secondary antibody against Swine IgG, heavy and light chains (whole IgG) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-25