Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-SWINE IGG (H+L) ANTIBODY  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52100173
Affinity purified polyclonal antibody to Swine IgG, both heavy chain and light chains made in Goat. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-668-15