Learn More
Seracare Life Sciences Inc ANTI-SWINE IGM (MU) AB AP
Supplier:  Seracare Life Sciences Inc 52200307
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Swine IgM, heavy chains (mu) made in Goat and labeled with alkaline phosphatase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-670-07