Learn More
Seracare Life Sciences Inc ANTI-SWINE IGM (MU) AB HRP
Supplier:  Seracare Life Sciences Inc 52200282
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Swine IgM, heavy chains (mu) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form.
Catalog No. 50-669-37