Learn More
ECM Bioscience Syk (Central region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience SM3731
Encompass_Preferred
Clone M373 was generated from a sequence corresponding to amino acids in the central region of human Syk. This sequence has high homology to mouse and rat Syk. Accession #P43405. This antibody detects a 72 kDa* protein corresponding to the molecular mass of Syk on SDS-PAGE immunoblots of adult mouse spleen. Applications: WB, E; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal, protein A purified antibody is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at -20°C. Stable for 1 year. Other Names =
Catalog No. 50-206-8853
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.