Learn More
Syk Mouse anti-Human, Clone: 4D10.1, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05434
Catalog No. 05-434-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect Syk using this Anti-Syk Antibody, clone 4D10.1 validated for use in immunoprecipitation, EA and western blotting.
Specifications
Specifications
Syk
4D10.1
Protein G Purified immunoglobulin in Protein A Purified immunoglobulin in 0.2M Tris-glycine, pH 7.2, 0.15M NaCl and 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
κ
100μg
Primary
Human
Immunoprecipitation, Western Blot
Unconjugated
NM_003177
SYK
Synthetic peptide corresponding to residues 313-339 of human Syk conjugated to KLH.
Protein A purfied
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Monoclonal