Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SYK, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132945

Catalog No. NBP132945


Add to cart

Description

Description

SYK Polyclonal antibody specifically detects SYK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

SYK
0.85mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
SYK
Recombinant protein encompassing a sequence within the center region of human SYK. The exact sequence is proprietary.
72 kDa
0.1 ml
Immunology, Innate Immunity, Mast Cell Markers, Protein Kinase
Primary
6850
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300
DKFZp313N1010, EC 2.7.10, EC 2.7.10.2, FLJ25043, spleen tyrosine kinaseFLJ37489, tyrosine-protein kinase SYK
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only