Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syk Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138694

Catalog No. 01-674-686


Add to cart

Description

Description

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is bone marrow or lymphoma.

SYK, a protein tyrosine kinase (PTK) is expressed in immature T cells, all B cells, basophils, hepatocytes, platelets, and erythrocytes. It functions in some T cell receptor signaling processes in addition to B cell receptor and integrin-mediated signal transduction processes. SYK-/SYK- mice exhibit defective phagocytosis by macrophages.
Specifications

Specifications

Syk
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
P43405
p72-Syk, tyrosine-protein kinase SYK
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6850
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SYK
Liquid
SYK
Synthetic peptide derived from internal region of human SYK protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit