Learn More
Synapsin 2 Sheep anti-Rat, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen OSS00072W
Catalog No. PIOSS00072W
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Glycerol (1:1) may be added for additional stability.
neuronal phosphoprotein that coats synaptic vesicles; binds to the cytoskeleton; may be important for vesicle trafficking to synapses and for the regulation of neurotransmitter release.
Specifications
Specifications
Synapsin 2
Polyclonal
whole serum with no preservative
Q63537
Synapsin 2, Syn2, Synapsin II
Sheep
IgG
RUO
Antibody
Polyclonal
Rat
Immunohistochemistry, Western Blot
Unconjugated
Syn2
Lyophilized
SYN2
A synthetic peptide from rat Synapsin-2
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
29179