Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synapsin 2 Sheep anti-Rat, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen OSS00073G

Catalog No. PIOSS00073G


Add to cart

Description

Description

Glycerol (1:1) may be added for additional stability. Reconstitute using 500 uL of sterile water.

neuronal phosphoprotein that coats synaptic vesicles; binds to the cytoskeleton; may be important for vesicle trafficking to synapses and for the regulation of neurotransmitter release.
Specifications

Specifications

Synapsin 2
Polyclonal
Unconjugated
Syn2
Lyophilized
SYN2
A synthetic peptide from rat Synapsin-2
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Rat
Immunohistochemistry, Western Blot
1 mg/mL
Antibody with no preservative
Q63537
Synapsin 2, Syn2, Synapsin II
Sheep
IgG
500 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
29179
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit