Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYNGR1 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA11044

Catalog No. PA11044


Add to cart

Description

Description

PA1-1044 detects Synaptogyrin 1 in rat and mouse samples. PA1-1044 has been successfully used in Western blot procedures. By Western blot, this antibody detects a ~25.5 kDa protein representing Synaptogyrin 1 in rat and mouse brain tissue samples. The PA1-1044 immunogen is a synthetic peptide corresponding to residues D(187) P S Q D S S M P Y A P Y V E P(202) of rat Synaptogyrin 1. This sequence is conserved in mouse and human. The PA1-1044 immunizing peptide (Cat. No. PEP-280) is available for use in neutralization and control experiments.

Synaptogyrins comprise a family of tyrosine-phosphorylated proteins with two neuronal (synaptogyrins 1 and 3) isoforms. Previous studies have indicated that synaptogyrins are involved in the regulation of neurotransmitter release. Synaptogyrin 1 is an integral membrane protein ubiquitously expressed in the nervous system. Synaptogyrin has four transmembrane domains and an extended cytoplasmic tail but it is not glycosylated. Synaptogyrin 1 is a tyrosine-phosphorylated by c-src and possibly other tyrosine kinases. The function of Synaptogyrin 1 is not well understood, but it may be a useful marker for nerve terminals and diagnosing neuroendocrine tumors.

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Specifications

SYNGR1
Polyclonal
Unconjugated
Syngr1
Liquid
SYNGR1
Synthetic peptide corresponding to residues D(187) P S Q D S S M P Y A P Y V E P(202) of rat Synaptogyrin 1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse, Rat
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
O55100, Q62876
p29
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
20972, 29205
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit