Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptojanin 1 Mouse, Unlabeled, Clone: 26, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612248

 View more versions of this product

Catalog No. BDB612248


Add to cart

Description

Description

Phosphoinositide turnover is a mechanism of intracellular signaling that involves phosphorylation of the inositol ring to produce signaling molecules that include phosphoinositol (PI)-3-P, PI-3,4-P2, PI-3,4,5-P3, PI-4-P, and PI-4,5-P2. These moleclules function in many signaling pathways to regulate events at cell membranes, such as endocytosis, exocytosis, and cell morphology. Synaptojanin 1 is a phosphoinositide phosphatase that selectively cleaves the 3-, 4-, and 5- phosphates from PIs. Two splice variants of synaptojanin 1 include a 145kDa form that is found in adult brain, and a 170kDa that is widely expressed in non- neuronal cells. The 145kDa form contains a Sac1 domain that has phosphoinositide phosphatase activity, a second phosphatase domain that has only PI-5 phosphatase activity (PI-5), and a proline-rich SH3-binding domain. The 170kDa form contains these domains, as well as a second SH3-binding consensus sequence. Synaptojanin binds many proteins implicated in endocytosis, including amphiphysins and endophilin. Thus, synaptojanin 1 may regulate endocytosis through modification of the interactions between SH3-domain proteins and PIs.

Western Blotting

Specifications

Specifications

Synaptojanin 1
26
Unconjugated
Affinity Purified
Rat Synaptojanin 1 aa. 1145-1259
50μg
Cell Biology
Primary
Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.