Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Synaptophysin, Clone: YE269, ™ Non-distribution product as customer accommodation.

Manufacturer:  MilliporeSigma™ 041019/DEL

Catalog No. 50-171-153

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Synaptophysin is a protein of the presynaptic vesicle exocytosis machinery interacting at the vesicle with VAMP/synaptobrevin. Synaptophysin structure consists of a hexameric protein of four transmembrane spans with cytoplasmic amino and carboxyl termini. Although its actual function remains elusive, synaptophysin remains a specific presynaptic marker for all neurons, neuroendocrine cells and neuroderived neoplasms.

Host Species: Rabbit
Clone: YE269
Isotype: IgG
Species Reactivity: Mouse, Rat, Human, Bovine
Epitope: C-terminus
Immunogen: Synthetic peptide corresponding to residues in C-terminus (cytoplasmic domain) of human Synaptophysin.
Formula Weight [Chemical]: ~34kDa

Immunocytochemistry, Immunohistochemistry, Western Blotting

Specifications

Specifications

Synaptophysin
Unconjugated
Rabbit
IgG
RUO
Primary
Bovine, Human, Murine, Rat
YE269
Unpurified
Synthetic peptide corresponding to residues in C-terminus (cytoplasmic domain) of human Synaptophysin.
100μL
Neuroscience
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit