Learn More
Synaptotagmin 1 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSS00022W
Catalog No. PIOSS00022W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
Synaptotagmin 1 is a synaptic vesicle membrane glycoprotein that is widely expressed throughout the CNS and is generally thought to act as the Ca2+ sensor in the regulation of exocytosis and neurotransmitter release. Studies indicate that synaptotagmin’s Ca2+ mediated binding of SNAP25 is essential for the Ca2+ dependent triggering of membrane. It has been demonstrated that discrete residues within the c(2)b binding domain of synaptotagmin 1 independently specify endocytic rate and synaptic vesicle size. Synaptotagmin 1 may have a regulatory role in the membrane interactions during trafficking of synaptic vesicles at the active zone of the synapse. Further, Synaptotagmin 1 binds acidic phospholipids with a specificity that requires the presence of both an acidic head group and a diacyl backbone. A Ca(2+)-dependent interaction between synaptotagmin and putative receptors for activated protein kinase C has been reported. Synaptotagmin 1 can also bind to at least three additional proteins in a Ca(2+)-independent manner; these are neurexins, syntaxin and AP2. Diseases associated with the dysfunction of Synaptotagmin 1 include food-borne botulism and infant botulism.
Specifications
Specifications
Synaptotagmin 1
Polyclonal
whole serum with no preservative
P46096, P21707
Synaptotagmin-1, Synaptotagmin I, SytI, p65, SVP65
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
Syt1
Lyophilized
SYT1
A synthetic peptide from n-terminal region of rat Synaptotagmin-1
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
20979, 25716