Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptotagmin 1 Sheep anti-Human, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen OSS00063G

Catalog No. PIOSS00063G


Add to cart

Description

Description

Glycerol (1:1) may be added for additional stability. Reconstitute in 500 µL of sterile water.

Synaptotagmin 1 is a synaptic vesicle membrane glycoprotein that is widely expressed throughout the CNS and is generally thought to act as the Ca2+ sensor in the regulation of exocytosis and neurotransmitter release. Studies indicate that synaptotagmin’s Ca2+ mediated binding of SNAP25 is essential for the Ca2+ dependent triggering of membrane. It has been demonstrated that discrete residues within the c(2)b binding domain of synaptotagmin 1 independently specify endocytic rate and synaptic vesicle size. Synaptotagmin 1 may have a regulatory role in the membrane interactions during trafficking of synaptic vesicles at the active zone of the synapse. Further, Synaptotagmin 1 binds acidic phospholipids with a specificity that requires the presence of both an acidic head group and a diacyl backbone. A Ca(2+)-dependent interaction between synaptotagmin and putative receptors for activated protein kinase C has been reported. Synaptotagmin 1 can also bind to at least three additional proteins in a Ca(2+)-independent manner; these are neurexins, syntaxin and AP2. Diseases associated with the dysfunction of Synaptotagmin 1 include food-borne botulism and infant botulism.
Specifications

Specifications

Synaptotagmin 1
Polyclonal
Unconjugated
SYT1
Lyophilized
SYT1
A synthetic peptide from human Synaptotagmin-1 (Synaptotagmin I) conjugated to an immunogenic carrier protein was used as the immunogen
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
1 mg/mL
Antibody with no preservative
P21579
Synaptotagmin-1, Synaptotagmin I, SytI, p65, SVP65
Sheep
IgG
500 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6857
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit