Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Synaptotagmin 2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSS00026W
Catalog No. PIOSS00026W
Description
Description
Reconstitute with 100 µL of distilled water.
Specifications
Specifications
Synaptotagmin 2
Polyclonal
whole serum with no preservative
Q8N9I0
Synaptotagmin II, SytII, SYT2
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
SYT2
Lyophilized
SYT2
A synthetic peptide from human Synaptotagmin-2
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
127833