Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Synaptotagmin 2, Polyclonal, Novus Biologicals™

Sheep Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130651

Catalog No. NBP130651


Add to cart

Description

Description

Synaptotagmin 2 Polyclonal antibody specifically detects Synaptotagmin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Synaptotagmin 2
LYOPH
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500
Purified
SYT2
A synthetic peptide from the n-terminal region of human Synaptotagmin-2 (Synaptotagmin II, SytII, SYT2) conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in rat and mouse.
IgG purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Appears to be specific for SYTII.
Human, Mouse, Rat
Immunohistochemistry, Western Blot
Unconjugated
Sterilized water with No Preservative
FLJ42519, synaptotagmin 2, synaptotagmin IISytII, synaptotagmin-2
Sheep
IgG
0.5 mg
Membrane Vesicle Markers
Primary
127833
Reconstitute with 0.5 ml sterilized water.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only