Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptotagmin 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific OSS00023W
Description
Reconstitute with 100 μL of distilled water.
Specifications
Synaptotagmin 2 | |
Polyclonal | |
SYT2 | |
Synaptotagmin II, SytII, SYT2 | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q8N9I0, P46097, P29101 | |
SYT2 | |
A synthetic peptide from the cytoplasmic domain of rat Synaptotagmin-2 | |
RUO | |
127833, 20980, 24805 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction