Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptotagmin Mouse, Unlabeled, Clone: 41, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610434

 View more versions of this product

Catalog No. BDB610434


Add to cart

Description

Description

Synaptotagmin (p65) is an abundant synaptic vesicle protein that contains a single transmembrane region and two copies of an internal repeat that is homologous to the regulatory region of Protein Kinase C. It appears that synaptotagmin has a regulatory role in the synaptic vesicle pathway, particularly in vesicle docking and/or fusion with the plasmalemma. A model has been proposed to explain docking, activation, and fusion of synaptic vesicles with donor membranes. This model suggests that VAMP/synaptobrevin and synaptotagmin (vSNARE) on the synaptic vesicle, and SNAP-25 and syntaxin (tSNAREs) on the plasma membrane, interact to form a 7S complex. Two additional soluble proteins, αSNAP and NSF, are later added to the 7S complex, accompanied by the loss of synaptotagmin. The resulting 20S complex contains syntaxin, SNAP-25, VAMP, αSNAP, and NSF. Genetic studies in several species demonstrate that mutation or deletion of synaptotagmin results in a large decrease in Ca2+ triggered transmitter release. Mammalian synapses that lack synaptotagmin show a selective decrease in a fast component of release, suggesting that synaptotagmin is the Ca2+ sensor triggering exocytosis.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

Synaptotagmin
41
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150μg
Cell Biology
Primary
Human, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.