Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptotagmin I Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB560050UL
Description
Detect Synaptotagmin I using this Anti-Synaptotagmin I Antibody, luminal part validated for use in western blot, immunohistochemistry.Specifications
Synaptotagmin I | |
Polyclonal | |
Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, 5% sucrose as a stabilizer and 0.025% sodium azide as a preservative. Reconstitute with 50μL of sterile deionized water. Centrifuge antibody prep | |
Rabbit | |
Affinity Purified | |
Primary | |
Rat |
Immunohistochemistry, Western Blot | |
Unconjugated | |
SYT1; DKFZp781D2042; SVP65; SytI; SYT; p65; P65; Synaptotagmin-1 | |
Peptide corresponding to amino acids 1-19 from rat Synaptotagmin I (Accession # P21107). | |
50 μL | |
NM_005639.1 | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction