Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Synaptotagmin (p Ser309), Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP14191450UG

Catalog No. NBP14191450

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Synaptotagmin (p Ser309)
1 mg/mL
Immunocytochemistry/Immunofluorescence 1:100-1:200
Affinity Purified
SYT3
synthesized phosphopeptide derived from human Synaptotagmin around the phosphorylation site of serine 309 (G-L-SP-D-P).
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence
Unconjugated
PBS (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
DKFZp761O132, synaptotagmin IIISytIII, synaptotagmin-3
Rabbit
IgG
50 ug
Membrane Vesicle Markers, Phospho Specific
Primary
84258
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only