Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptotagmin V Mouse, Unlabeled, Clone: 46, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612284

Catalog No. BDB612284


Add to cart

Description

Description

Synaptotagmin I belongs to a family of C2 domain containing proteins, which include Synatotagmin II to XI and Srg1. All of the members of this protein family contain a single transmembrane region and two copies of an internal repeat that is homologous to the regulatory region of Protein Kinase C (C2A and C2B domains). Synaptotagmin I-V and IX-XI are found in brain, while synaptotagmin VI-VIII are found in non-neuronal tissues. Synaptotagmin I and II have regulatory roles in synaptic vesicle docking and fusion with the plasmalemma. Both synaptotagmin I and II act as vSNAREs that form a complex with SNAP-25 and syntaxin at the plasma membrane. This complex is required for rapid, calcium-dependent synaptic vesicle exocytosis. In addition to their roles in exocytosis, synaptotagmin I and II can bind to calcium channels, calmodulin, and the clathrin adaptor protein AP-2. Synatotagmins III, V, VI, and X cannot bind IP4 like other synaptotagmins, but like most synaptotagmins these family members form heteromers with other synaptotagmins. Synaptotagmins IV, X, and Srg1 may regulate vesicular trafficking during brain development and plasticity. In neuronal and non-neuronal tissues, synaptotagmins may be important for vesicle exocytosis, endocytosis, and trafficking, as well calcium signaling.

Western Blotting

Specifications

Specifications

Synaptotagmin V
46
Unconjugated
Affinity Purified
Rat Synaptotagmin V aa. 15-110
50μg
Cell Biology
Primary
Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit