Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYNC Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524290

Catalog No. PIPA524290

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a member of the intermediate filament family which contains an N-terminal head domain, followed by a central coiled-coil region and a short C-terminal tail. The protein is highly expressed in skeletal and cardiac muscle. The protein links the dystrophin associated protein complex (DAPC) to desmin filaments in muscle and may have a structural role in striated muscle. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

SYNC
Polyclonal
PBS with 0.09% sodium azide
Q9H7C4
SYNC1, SYNCOILIN, intermediate filament protein syncoilin, syncoilin, syncoilin intermediate filament 1, syncoilin, intermediate filament 1, syncoilin-1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
81493
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
SYNC
Liquid
SYNC
KLH conjugated synthetic peptide between 129-158 amino acids from the N-terminal region of human SYNCI
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit