Learn More
Syndecan 4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA132485
Description
PA1-32485 detects Syndecan 4 from mouse, rat, human samples. PA1-32485 has been successfully used in immunohistochemistry (frozen tissue), immunohistochemistry (paraffin tissue), Western blot, immunoprecipitation applications. Western blot detects a band of approximately 24kDa (predicted molecular weight: 22kDa. The PA1-32485 immunogen is synthetic peptide surrounding amino acid 184 of human Syndecan 4. Store at 4°C short term. For extended storage aliquot and store at -20°C or below. Avoid freeze/thaw cycles.
Syndecans are tyupe I integral membrane proteoglycans that are involved in cell-extracellular matrix adhesion and growth factor binding. Syndecan 1 is a matrix receptor which binds to Collagens, Fibronectin and Thrombospondin. Syndecan 1 and Syndecan 3 interact with MK (midkine), a growth/differentiation factor involved in embryogenesis. Syndecan 2 is highly expressed at areas of high morphogenetic activity, such as epithelial-mesencymal interfaces and the prechondrogenic and preosteogenic mesenchymal condensations. Syndecan 4 functions cooperatively with integrins in the processes of cell spreading and focal adhesion assembly.Specifications
Syndecan 4 | |
Polyclonal | |
Unconjugated | |
P31431, O35988, P34901 | |
SDC4 | |
Synthetic peptide surrounding amino acid 184 of human Syndecan 4. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
SDC4 | |
MGC22217, SYND4 | |
Rabbit | |
Protein A | |
RUO | |
20971, 24771, 6385 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.