Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD138, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA121165

Catalog No. PA121165

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CD138 / Syndecan-1 is a transmembrane heparin sulphate proteoglycan which is made up of one core protein and five glycosaminoglycan. CD138 is expected to play a role in cell adhesion. It is expressed on the surface of pre-B cells and plasma cells but is absent from mature B cells. It is a selective marker for B cell lymphoblastic leukemia and lymphoplasmocytoid leukemia. It is lost from the apoptotic myeloma cells; hence is a useful marker for viable myeloma cells.

PA1-21165 detects Syndecan in Human samples.PA1-21165 has been successfully used in Immunohistochemistry (Paraffin) procedures. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citratee buffer, pH 6.0 for 10 minutes followed by cooling at room temperature for 20 minutes.PA1-21165 immunogen corresponds to Recombinant fragment, corresponding to N terminal amino acids 18-218 of Human Syndecan..
Specifications

Specifications

Syndecan 1/CD138
Polyclonal
Unconjugated
SDC1
Purified
SDC1
Recombinant fragment, corresponding to N terminal amino acids 18-218 of Human Syndecan.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
0.2mg/mL
PBS with 1% BSA and 0.09% sodium azide; pH 7.4
P18827
SDC1, SYND1, SDC, CD antigen 138
Rabbit
IgG
500μL
4° C, do not freeze, do not freeze
Primary
6382
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit