Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
spectrin repeat containing, nuclear envelope 1, Mouse, Clone: 4C3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SYNE1.
Supplier: Abnova Corporation H00023345M01
Description
This gene encodes a spectrin repeat containing protein expressed in skeletal and smooth muscle, and peripheral blood lymphocytes, that localizes to the nuclear membrane. Mutations in this gene have been associated with autosomal recessive spinocerebellar ataxia 8, also referred to as autosomal recessive cerebellar ataxia type 1 or recessive ataxia of Beauce. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq
Sequence: LRKIQQSVSEFEDKLAVPIKICSSATETYKVLQEHMDLCQALESLSSAITAFSASARKVVNRDSCVQEAAALQQQYEDILRRAKERQTALENLLAHWQRLEKELSSFLTWSpecifications
spectrin repeat containing, nuclear envelope 1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_182961 | |
SYNE1 | |
SYNE1 (NP_892006, 1561 a.a. ∼ 1670 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
50 μg | |
Primary | |
Human | |
Antibody |
ELISA, Western Blot | |
4C3 | |
Mouse monoclonal antibody raised against a partial recombinant SYNE1. | |
SYNE1 | |
8B/CPG2/DKFZp781J13156/FLJ30878/FLJ41140/KIAA0796/KIAA1262/KIAA1756/MYNE1/SCAR8 | |
Mouse | |
Affinity Purified | |
RUO | |
23345 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG3 κ |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction