Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYNE3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524310

Catalog No. PIPA524310


Add to cart

Description

Description

Component of SUN-protein-containing multivariate complexes also called LINC complexes which link the nucleoskeleton and cytoskeleton by providing versatile outer nuclear membrane attachment sites for cytoskeletal filaments. Involved in the maintece of nuclear organization and structural integrity. Probable anchoring protein which tethers the nucleus to the cytoskeleton by binding PLEC which can associate with the intermediate filament system.
Specifications

Specifications

SYNE3
Polyclonal
Unconjugated
SYNE3
Liquid
SYNE3
KLH conjugated synthetic peptide between 852-880 amino acids from the C-terminal region of human SYNE3
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.45 mg/mL
PBS with 0.09% sodium azide
Q6ZMZ3, Q4FZC9
NET53, SYNE3, nesprin-3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
161176, 212073
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit