Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYNGR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA520879

Catalog No. PIPA520879


Add to cart

Description

Description

A suggested positive control is K562 cell lysate. PA5-20879 can be used with blocking peptide PEP-0993.

Synaptogyrins comprise a family of tyrosine-phosphorylated membrane proteins with two neuronal (SYNGR1 and SYNGR3) and one ubiquitous (SYNGR2) members. SYNGR1 and -3 are synaptic vesicle proteins, residing in some cases on the same synaptic vesicle, and are thought to be involved in the regulation of neurotransmitter release. SYNGR2, by contrast, is absent from synaptic vesicles. SYNGR3 is predomitly expressed in brain and placenta. The role and localization of a fourth synaptogyrin, SYNGR4, is unclear.
Specifications

Specifications

SYNGR4
Polyclonal
Unconjugated
SYNGR4
Liquid
SYNGR4
A 14 amino acid peptide from near the amino terminus of human SYNGR4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O95473, Q9Z1L2
synaptogyrin-4
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
23546, 292916, 58867
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit