Learn More
Synphilin 1 Goat anti-Human, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Manufacturer:  Invitrogen PA184168
Catalog No. PIPA184168
Add to cart
Description
Description
A suggested positive control for immunohistochemical applications is human brain.
This gene encodes a protein containing several protein-protein interaction domains, including ankyrin-like repeats, a coiled-coil domain, and an ATP/GTP-binding motif. The encoded protein interacts with alpha-synuclein in neuronal tissue and may play a role in the formation of cytoplasmic inclusions and neurodegeneration. A mutation in this gene has been associated with ParkinsoNULLs disease. Alternatively spliced transcript variants encoding different isoforms of this gene have been described, but their full-length nature has yet to be determined.
Specifications
Specifications
Synphilin 1
Polyclonal
whole serum with 0.1% sodium azide
Q9Y6H5
MGC39814, SYPH1
Goat
IgG
RUO
Antibody
Polyclonal
Human
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SNCAIP
Liquid
SNCAIP
Peptide 829SLELNGEKDKDKGRTLQRT847.
100 μL
4°C or -20°C if preferred
Primary
9627