Learn More
Synphilin 1 Rabbit anti-Human, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA118269
Catalog No. 01-670-522
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-18269 detects Synphilin-1 from human and rat samples. PA1-18269 has been successfully used in immunohistochemistry procedures. The PA1-18269 immunogen is a synthetic peptide corresponding to residues S L E L N G E K D K D K G R T L Q R T of human synphilin-1a, conjugated to KLH. Reconstitute with 100 μL of distilled water.
Synuclein alpha interacting protein (Synphilin-1) contains several protein-protein interaction domains and interacts with alpha synuclein in neurons. Mutations of SNCAIP have been linked to Parkinson disease. The amino acid sequence of synphilin-1 shares a high level of identity with its human counterpart, particularly in regions containing ankyrin-like motifs and the coiled-coil domain. Expression pattern of synphilin-1 in tissues is similar in both mouse and human. Synphilin-1 has an important role in the formation of aggregates and cytotoxicity in Parkinson disease and also Dorfin may be involved in the pathogenic process by ubiquitylation of synphilin-1.
Specifications
Specifications
Synphilin 1
Polyclonal
whole serum with no preservative
Q9Y6H5
Alpha-synuclein-interacting protein, SNCAIP
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Rat
Immunohistochemistry
Unconjugated
SNCAIP
Lyophilized
SNCAIP
Synthetic peptide corresponding to residues S L E L N G E K D K D K G R T L Q R T of human synphilin-1a.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
307309, 9627