Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntaxin 6 Mouse, Unlabeled, Clone: 30, BD

Mouse Monoclonal Antibody

$225.00 - $445.00

Specifications

Antigen Syntaxin 6
Isotype IgG1
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610635
View Documents
BD Biosciences
610635
50μg Each for $225.00
Add to cart
 
BDB610636
View Documents
BD Biosciences
610636
150μg Each for $445.00
Add to cart
 
Description

Description

Signal transmission between cells is often regulated by the secretion of neurotransmitters or hormones into the extracellular space. This process is controlled by a complex pathway of membrane trafficking which leads to membrane fusion and secretion. Syntaxin 1 is involved in three important complexes that modulate the secretion process: syntaxin and n-sec-1, syntaxin, VAMP, and SNAP-25, and syntaxin, VAMP, SNAP-25, αSNAP, and NSF (20S complex). Syntaxin 6 is a 255 amino acid member of the syntaxin family. It contains a C-terminal transmembrane domain and is located at the Golgi apparatus. In addition, Syntaxin 6 displays significant (47% identity) homology with SNAP-25. In vitro, Syntaxin 6 has been reported to interact with α-SNAP. Syntaxin 6 has been reported to have a calculated molecular weight of 30.6kDa, but has been observed to migrate in a range inbetween ~ 31-35kDa.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Syntaxin 6
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Unconjugated
Cell Biology
Murine
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
30
250μg/mL

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit