Learn More
STX7 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSS00030W
Catalog No. PIOSS00030W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
Specifications
Specifications
STX7
Polyclonal
whole serum with no preservative
O15400
Syntaxin-7, STX7, Syn7
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
STX7
Lyophilized
STX7
A synthetic peptide from human Syntaxin-7
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
8417