Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STX7 Sheep anti-Human, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Supplier:  Thermo Scientific OSS00057G

Catalog No. PIOSS00057G

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Glycerol (1:1) may be added for additional stability.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

STX7
Polyclonal
Unconjugated
O15400
STX7
A synthetic peptide from human Syntaxin-7 (STX7, Syn7, Syntaxin 7) conjugated to an immunogenic carrier protein was used as the immunogen
500 μg
Primary
Human
Antibody
IgG
Immunohistochemistry, Western Blot
1 mg/mL
STX7
Syntaxin-7, STX7, Syn7
Sheep
Ammonium sulfate precipitation
RUO
8417
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit