Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntaxin 8 Mouse, Unlabeled, Clone: 48, BD

Mouse Monoclonal Antibody

$207.00 - $419.00

Specifications

Antigen Syntaxin 8
Isotype IgG2a
Immunogen Mouse Syntaxin 8 aa. 57-164
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB611352
View Documents
BD Biosciences
611352
50μg Each for $207.00
Add to cart
 
BDB611353
View Documents
BD Biosciences
611353
150μg Each for $419.00
Add to cart
 
Description

Description

Eukaryotic protein trafficking involves the packaging of molecules into membranous vesicles that bud from a donor compartment, travel to a specific destination, fuse, and release their components into an acceptor compartment. Recognition between vesicle and acceptor membrane is mediated by the pairing of the integral membrane SNARE proteins. The stable interaction between vesicle proteins (v-SNAREs) and target proteins (t-SNAREs) juxtaposes the membranes and results in an activated docking state and/or membrane fusion. The syntaxin protein family contains a number of members that serve as functional t-SNAREs. One member of this family, syntaxin 8, has two coiled-coil domains, one in the N-terminus and one toward the C-terminus. The C-terminal domain is homologous to a similar domain in syntaxin 6 and is predicted to interact in the formation of SNARE complexes, while a hydrophobic domain in the C-terminus may be involved in membrane anchoring. Syntaxin-8 is ubiquitously expressed, but is present at high levels in the heart and localizes specifically to the ER. Thus, syntaxin-8 is a t-SNARE that is thought to be involved in the early secretory pathway of many different cell types.

Immunofluorescence, Western Blotting

Specifications

Specifications

Syntaxin 8
Mouse Syntaxin 8 aa. 57-164
Unconjugated
Cell Biology
Murine
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG2a
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
48
250μg/mL

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit