Learn More
MilliporeSigma™ anti-Synuclein, α Ab-4; Polyclonal
Supplier: MilliporeSigma™ AB5038/DEL
Description
α -Synuclein is a small (140-amino acid) cytoplasmic protein of unclear function that is enriched in presynaptic terminals and is the precursor protein of a nonamyloid β component of senile plaques in Alzeheimer's Disease (AD). α -Synuclein is located predominantly in the presynaptic nerve terminals in the brain but is also found in low concentrations in all tissues except liver. Recent studies have shown that abnormal aggregation and accumulation of synaptic proteins, including alpha-Synuclein might be associated with plaque formation in AD and seems to be a major component of Lewy bodies in dementia with Lewy bodies (DLB). It has also been identified in the Glial Cytoplasmic Inclusions (GCIs) found in Multiple System Atrophy (MSA).
Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting
Specifications
Synuclein, α Ab-4 | |
Unconjugated | |
Human α-synuclein residues 111-131 coupled to KLH. The peptide sHuence is N-GILEDMPVDPDNEAYEMPSEEC-C. | |
RUO | |
Primary | |
Serum |
Polyclonal | |
Rabbit | |
50 μL | |
Neuroscience | |
Human, Rat |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.