Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Synuclein-gamma, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP104361
Description
gamma-Synuclein Polyclonal specifically detects gamma-Synuclein in Human samples. It is validated for Western Blot, ELISA.Specifications
gamma-Synuclein | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
BCSG1PERSYN, Breast cancer-specific gene 1 protein, gamma-synuclein, persyn, SRPRSN, synoretin, synuclein, gamma (breast cancer-specific protein 1) | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
This is specific for gamma-synuclein and does not cross-react with alpha- and beta-synuclein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 1:500-1:2000, ELISA 1:100-1:2000 | |
NP_003078.2 | |
SNCG | |
Recombinant human gamma-Synuclein purified from E. coli | |
0.1 mL | |
Alzheimers Research, Mitotic Regulators, Neuroscience | |
6623 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction