Learn More
anti-Synuclein-gamma, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP131209
Catalog No. NBP131209
Add to cart
Description
Description
gamma-Synuclein Polyclonal antibody specifically detects gamma-Synuclein in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
gamma-Synuclein
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1-100-1:1000, Immunohistochemistry-Frozen
Affinity Purified
SNCG
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human gamma Synuclein. The exact sequence is proprietary.
13 kDa
100 ul
Alzheimers Research, Cellular Markers, Mitotic Regulators, Neuroscience
Primary
6623
Human, Mouse
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
BCSG1PERSYN, Breast cancer-specific gene 1 protein, gamma-synuclein, persyn, SRPRSN, synoretin, synuclein, gamma (breast cancer-specific protein 1)
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). Not yet tested in any other species.