Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYTL3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524291

Catalog No. PIPA524291

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May act as Rab effector protein and play a role in vesicle trafficking. Binds phospholipids in the presence of calcium ions.
Specifications

Specifications

SYTL3
Polyclonal
PBS with 0.09% sodium azide
Q4VX76, Q99N48
SLP3, exophilin-6, synaptotagmin-like protein 3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
83672, 94120
Western Blot
Unconjugated
SYTL3
Liquid
SYTL3
KLH conjugated synthetic peptide between 106-135 amino acids from the N-terminal region of human SYTL3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit