Learn More
T-Cell Receptor Mouse anti-Human, Clone: C305, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05919
Catalog No. 05-919-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-T-Cell Receptor Antibody, clone C305 detects level of T-Cell Receptor and has been published and validated for use in Mplex and immunoprecipitation.
Specifications
Specifications
T-Cell Receptor
C305
Supernatant
Murine
IgM
6μg
Primary
Human
Immunoprecipitation
Unconjugated
TRA@, TCRD, FLJ22602, MGC22624, TRA, TRDD3, MGC71411, MGC117436, MGC23964, TCRA
Jurkat T cells
Concentrated culture supernatant
−20°C for up to 24 months.
Monoclonal